3,5-Di-tert-butyl-4-hydroxybenzylalkohol ≥95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR2794-5G
OR2794-25G
OR2794-100G
APOSOR2794-5GEA
27.7
EUR
APOSOR2794-5G
APOSOR2794-25G
APOSOR2794-100G
3,5-Di-tert-butyl-4-hydroxybenzylalkohol ≥95%
3,5-Di-tert-butyl-4-hydroxybenzylalkohol
Formel:
C₁₅H₂₄O₂ Molecular Weight: 236.35 g/mol Siedepunkt: 138…141 °C Schmelzpunkt: 214 °C |
MDL:
MFCD00017254 CAS-Nummer: 88-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...