4-Amino-2-cyclopropylbenzoic acid ≥95%
Lieferant: Apollo Scientific
OR70329-250MG OR70329-1G OR70329-5G
APOSOR70329-250MGEA 190 EUR
APOSOR70329-250MG APOSOR70329-1G APOSOR70329-5G
4-Amino-2-cyclopropylbenzoic acid ≥95%
4-Amino-2-cyclopropylbenzoic acid
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.19984 g/mol
MDL: MFCD32862615
CAS-Nummer: 2609110-47-4