3-Bromo-5-cyclopropylbenzoic acid ≥95%
Lieferant: Apollo Scientific
OR70321-250MG
OR70321-1G
APOSOR70321-250MGEA
219
EUR
APOSOR70321-250MG
APOSOR70321-1G
3-Bromo-5-cyclopropylbenzoic acid ≥95%
3-Bromo-5-cyclopropylbenzoic acid
Formel:
C₁₀H₉BrO₂ Molecular Weight: 241.08126 g/mol |
MDL:
MFCD26959998 CAS-Nummer: 1211583-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...