2-Bromo-3-chloro-5-iodobenzaldehyde 95%
Lieferant: Apollo Scientific
OR61547-250MG
OR61547-1G
APOSOR61547-250MGEA
190
EUR
APOSOR61547-250MG
APOSOR61547-1G
2-Bromo-3-chloro-5-iodobenzaldehyde 95%
2-Bromo-3-chloro-5-iodobenzaldehyde
Formel:
C₇H₃BrClIO Molecular Weight: 345,35959 g/mol |
CAS-Nummer:
2090983-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...