2-Iodo-5-methoxybenzaldehyde ≥95%

Lieferant: Apollo Scientific

OR62152-1G OR62152-5G
APOSOR62152-1GEA 203 EUR
APOSOR62152-1G APOSOR62152-5G
2-Iodo-5-methoxybenzaldehyde ≥95%
2-Iodo-5-methoxybenzaldehyde
Formel: C₈H₇IO₂
Molecular Weight: 262.046 g/mol
Schmelzpunkt: 110…115 °C
MDL: MFCD18905098
CAS-Nummer: 77287-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR