5-Iodo-2-methyl-2H-pyrazolo[3,4-b]pyridine ≥95%

Lieferant: Apollo Scientific

OR62133-250MG OR62133-1G OR62133-5G
APOSOR62133-250MGEA 224 EUR
APOSOR62133-250MG APOSOR62133-1G APOSOR62133-5G
5-Iodo-2-methyl-2H-pyrazolo[3,4-b]pyridine ≥95%
5-Iodo-2-methyl-2H-pyrazolo[3,4-b]pyridine
Formel: C₇H₆IN₃
Molecular Weight: 259.0471 g/mol
MDL: MFCD32903685
CAS-Nummer: 2594436-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR