Methyl 3-fluoro-4-hydroxy-5-nitrobenzoate ≥95%

Lieferant: Apollo Scientific

PC502068-250MG PC502068-1G PC502068-5G
APOSPC502068-250MGEA 190 EUR
APOSPC502068-250MG APOSPC502068-1G APOSPC502068-5G
Methyl 3-fluoro-4-hydroxy-5-nitrobenzoate ≥95%
Methyl 3-fluoro-4-hydroxy-5-nitrobenzoate
Formel: C₈H₆FNO₅
Molecular Weight: 215.13534 g/mol
MDL: MFCD16038819
CAS-Nummer: 850335-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR