Methyl 3-amino-5-fluoro-4-hydroxybenzoate ≥95%
Lieferant: Apollo Scientific
PC502069-250MG
PC502069-1G
PC502069-5G
APOSPC502069-250MGEA
203
EUR
APOSPC502069-250MG
APOSPC502069-1G
APOSPC502069-5G
Methyl 3-amino-5-fluoro-4-hydroxybenzoate ≥95%
Methyl 3-amino-5-fluoro-4-hydroxybenzoate
Formel:
C₈H₈FNO₃ Molecular Weight: 185,15242 g/mol |
MDL:
MFCD20725712 CAS-Nummer: 946121-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...