1-(4-Bromo-2,3,6-trifluorophenyl)ethan-1-one ≥95%
Lieferant: Apollo Scientific
PC50548-250MG
PC50548-1G
APOSPC50548-250MGEA
219
EUR
APOSPC50548-250MG
APOSPC50548-1G
1-(4-Bromo-2,3,6-trifluorophenyl)ethan-1-one ≥95%
1-(4-Bromo-2,3,6-trifluorophenyl)ethan-1-one
Formel:
C₈H₄BrF₃O Molecular Weight: 253,01596 g/mol |
MDL:
MFCD32669844 CAS-Nummer: 1523271-90-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)