7-Bromo-4-fluoro-1,2-dihydroisoquinolin-1-one ≥95%

Lieferant: Apollo Scientific

PC55912-250MG PC55912-1G
APOSPC55912-250MGEA 400 EUR
APOSPC55912-250MG APOSPC55912-1G
7-Bromo-4-fluoro-1,2-dihydroisoquinolin-1-one ≥95%
7-Bromo-4-fluoro-1,2-dihydroisoquinolin-1-one
Formel: C₉H₅BrFNO
MDL: MFCD20526591
CAS-Nummer: 956700-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR