5-Bromo-2-(difluoromethyl)benzaldehyde ≥95%

Lieferant: Apollo Scientific

PC57735-250MG PC57735-1G PC57735-5G
APOSPC57735-250MGEA 190 EUR
APOSPC57735-250MG APOSPC57735-1G APOSPC57735-5G
5-Bromo-2-(difluoromethyl)benzaldehyde ≥95%
5-Bromo-2-(difluoromethyl)benzaldehyde
Formel: C₈H₅BrF₂O
MDL: MFCD28534225
CAS-Nummer: 1779823-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR