2-Brom-4-(trifluormethoxy)benzaldehyd ≥95%
Lieferant: Apollo Scientific
PC901498-1G
PC901498-5G
PC901498-10G
APOSPC901498-1GEA
125
EUR
APOSPC901498-1G
APOSPC901498-5G
APOSPC901498-10G
2-Brom-4-(trifluormethoxy)benzaldehyd ≥95%
2-Bromo-4-(trifluoromethoxy)benzaldehyde
Formel:
C₈H₄BrF₃O₂ |
MDL:
MFCD09835093 CAS-Nummer: 1114808-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...