3-Bromo-2-(bromomethyl)benzoic acid ≥95%
Lieferant: Apollo Scientific
OR61455-250MG
OR61455-1G
OR61455-5G
APOSOR61455-250MGEA
94.4
EUR
APOSOR61455-250MG
APOSOR61455-1G
APOSOR61455-5G
3-Bromo-2-(bromomethyl)benzoic acid ≥95%
3-Bromo-2-(bromomethyl)benzoic acid
Formel:
C₈H₆Br₂O₂ Molecular Weight: 293.94003 g/mol |
MDL:
MFCD11847530 CAS-Nummer: 1194354-14-7 UN: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...