5-Bromo-2-(difluoromethyl)benzonitrile ≥95%

Lieferant: Apollo Scientific

PC57762-250MG PC57762-1G
APOSPC57762-250MGEA 261 EUR
APOSPC57762-250MG APOSPC57762-1G
5-Bromo-2-(difluoromethyl)benzonitrile ≥95%
5-Bromo-2-(difluoromethyl)benzonitrile
Formel: C₈H₄BrF₂N
MDL: MFCD18391066
CAS-Nummer: 1128269-99-7
UN: 3439

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR