5-Bromo-2-(difluoromethyl)benzyl bromide ≥95%
Lieferant: Apollo Scientific
PC57771-250MG
PC57771-1G
APOSPC57771-250MGEA
219
EUR
APOSPC57771-250MG
APOSPC57771-1G
5-Bromo-2-(difluoromethyl)benzyl bromide ≥95%
5-Bromo-2-(difluoromethyl)benzyl bromide
Formel:
C₈H₆Br₂F₂ Molecular Weight: 299.93804 g/mol |
MDL:
MFCD18391076 CAS-Nummer: 1261760-28-4 UN: 3265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...