4-Bromo-2,5-bis(trifluoromethyl)benzonitrile ≥95%

Lieferant: Apollo Scientific

PC56691-250MG PC56691-1G
APOSPC56691-250MGEA 261 EUR
APOSPC56691-250MG APOSPC56691-1G
4-Bromo-2,5-bis(trifluoromethyl)benzonitrile ≥95%
4-Bromo-2,5-bis(trifluoromethyl)benzonitrile
Formel: C₉H₂BrF₆N
MDL: MFCD28727944
CAS-Nummer: 1805971-70-3
UN: 3276

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR