2,4-Dibromo-6-fluorobenzonitrile ≥95%
Lieferant: Apollo Scientific
PC57414-250MG
PC57414-1G
PC57414-5G
APOSPC57414-250MGEA
203
EUR
APOSPC57414-250MG
APOSPC57414-1G
APOSPC57414-5G
2,4-Dibromo-6-fluorobenzonitrile ≥95%
2,4-Dibromo-6-fluorobenzonitrile
Formel:
C₇H₂Br₂FN |
MDL:
MFCD22377850 CAS-Nummer: 1393585-66-4 UN: 3439 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...