CHOgro® Expression Media
Lieferant: MIRUS
MIR6200 MIR6201 MIR6202
MIRUMIR6200EA 0 EUR
MIRUMIR6200 MIRUMIR6201 MIRUMIR6202
CHOgro® Expression Media
Zelltrennmedien
CHOgro® Expression medium is formulated to provide high density cell growth.
  • Chemically defined
  • Hydrolysate-free
  • Grow many suspension CHO cells with minimal adaptation
CHOgro® Expression medium is a chemically defined, hydrolysate-free and animal-origin-free medium manufactured under cGMP conditions in an ISO-compliant facility. CHOgro® is formulated to provide high density cell growth, and many suspension CHO cells (e.g. Freestyle™ CHO-S) can easily and quickly grow in CHOgro® Medium with minimal adaptation.