TPCA-1 ≥98%

Lieferant: Cayman Chemical

Synonyme: 5-(4-Fluorophenyl)-2-ureidothiophene-3-carboxamide, 2-[(aminocarbonyl)amino]-5-(4-fluorophenyl)-3-thiophenecarboxamide, IKK2 Inhibitor IV

15115-10 15115-1 15115-5 15115-500
CAYM15115-10EA 267 EUR
CAYM15115-10 CAYM15115-1 CAYM15115-5 CAYM15115-500
TPCA-1 ≥98%
TPCA-1

  • A selective inhibitor of IKK2


A selective inhibitor of IKK2 (IC₅₀s = 17,9 nM and 0,40 μM for IKK2 and IKK1, respectively); reduces the severity and onset of collagen-induced arthritis in mice and blocks cytokine release in an antigen-driven rat model of lung inflammation.

Formel: C₁₂H₁₀FN₃O₂S
Molecular Weight: 279,3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09037541
CAS-Nummer: 507475-17-4

Bestellen

Spezifikation Testergebnisse

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR