NF-κB Inhibitor (SN50)
Lieferant: Cayman Chemical
17493-1
17493-5
17493-500
CAYM17493-1EA
170
EUR
CAYM17493-1
CAYM17493-5
CAYM17493-500
NF-κB Inhibitor (SN50)
Proteine und Peptide
A synthetic peptide corresponding to the nuclear localisation sequence of NF-κB p105 subunit (also known as p50) appended to a hydrophobic sequence to facilitate import into living cells; blocks the nuclear import of p105 in cells treated with activators of NF-κB signaling, including LPS and TNF-α.
- A cell-permeable peptide that blocks nuclear import of NF-κB
Stability: ≥2 years