Anti-Haemophilus influenza B Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)

Lieferant: Bioss
BOSSBS-15403R-A555EA 858 EUR
BOSSBS-15403R-A555
Anti-Haemophilus influenza B Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Antikörper
Haemophilus influenza B.

Type: Primary
Antigen: Haemophilus influenza B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 101-200/339
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR