Anti-ROCK2 Thr249 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BOSSBS-5675R-A750EA 885 EUR
BOSSBS-5675R-A750
Anti-ROCK2 Thr249 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The protein encoded by this gene is a serine/threonine kinase that regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibres and focal adhesions, and the activation of the c-fos serum response element. This protein, which is an isozyme of ROCK1 is a target for the small GTPase Rho.

Type: Primary
Antigen: ROCK2 Thr249
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR