Anti-PauA Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BOSSBS-9846R-A750EA 858 EUR
BOSSBS-9846R-A750
Anti-PauA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Plasminogen activator A (PauA) is a Streptococcus uberis protein that functions similarly to proteins within the Staphylokinase/Streptokinase family. Streptococcus uberis is a common cause of infection and inflammatory disease in the bovine udder. Inflammation of breast tissue (mastitis) has been recognised as a serious threat to the welfare of the dairy cow and has a severe economic impact on the dairy industry. S. uberis has been shown to specifically activate bovine and ovine plasminogen and this is mediated by the extracellular protein, plasminogen activator uberis (PauA), which has a molecular mass of around 30 kDa. It has been postulated that activation of plasminogen by PauA may facilitate early bacterial colonization of the lactating gland by promoting the release of nutrients which are likely to be restricted in milk.

Type: Primary
Antigen: PauA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 255-350/2876
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR