Anti-Heatstable Enterotoxin 1 [Escherichia coli] Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BOSSBS-8858R-A680EA 858 EUR
BOSSBS-8858R-A680
Anti-Heatstable Enterotoxin 1 [Escherichia coli] Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Heat-stable enterotoxins (STs) are secretory peptides produced by some bacterial strains of Escherichia coli.

Type: Primary
Antigen: Heatstable Enterotoxin 1 [Escherichia coli]
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 3-38/38
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR