(R)-(+)-Limonene, für die Nahrungsmittelindustrie
Lieferant: Spectrum Chemical
|
|
Synonyme:
(R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexen, (R)-(+)-Dipenten, (R)-(+)-p-Mentha-1,8-dien, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexen, D(+)-Limonen
D-Limonene, Food Grade, also known as 4-Isopropenyl-1-methylcyclohexene, is used as a flavorant and as a fragrance in perfumery, aftershave lotions, bath products and other such products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
GHS hazard : Flame;Health hazard;Exclamation mark
UN2052
Formel:
C₁₀H₁₆ Molecular Weight: 136.24 g/mol Siedepunkt: 175.5 Schmelzpunkt: -74 Dichte: 0,841 Flammpunkt: 43 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062991 CAS-Nummer: 5989-27-5 UN: 2052 ADR: 3 III Merck Index: 13,05515 |
Spezifikation Testergebnisse
Appearance | Transparent Liquid |
Assay (GC) | 95.0 - 97.5% |
Specific Gravity @ 25°C | 0.838 - 0.843 |
Refractive Index @ 20°C | 1.471 - 1.474 |
Optical Rotation @ 25°C | +96.0° to +104 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...