Choline chloride USP Nahrungsergänzung 
Lieferant: Spectrum Chemical
Synonyme: (2-Hydroxyethyl)trimethylammoniumchlorid
C2058-2.5KG C2058-500GM
SPCMC2058-2.5KGEA 748 EUR
SPCMC2058-2.5KG SPCMC2058-500GM
Choline chloride USP Nahrungsergänzung 
Cholinchlorid
Choline Chloride, USP Dietary Supplement.
All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formel: HOH₂CCH₂N(CH₃)₃Cl
Molecular Weight: 139.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011721
CAS-Nummer: 67-48-1
Merck Index: 13,02226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM