D-(-)-Sorbitol solution_SC 70% aqueous solution 70 USP
Lieferant: Spectrum Chemical
Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol
SO220-20LTPL SO220-500MLGL
SPCMSO220-20LTPLEA 976 EUR
SPCMSO220-20LTPL SPCMSO220-500MLGL
D-(-)-Sorbitol solution_SC 70% aqueous solution 70 USP
D(-)-Sorbit
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Dichte: 1,280
Lagertemperatur: Raumtemperatur
MDL: MFCD00004708
CAS-Nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM