Allantoin 98,5 USP
Lieferant: Spectrum Chemical
Synonyme:
1-(2,5-Dioxoimidazolidin-4-yl)urea
A2721-25KGBL
A2721-50KGBL
A2721-500GM
A2721-2.5KG
SPCMA2721-25KGBLEA
8090
EUR
SPCMA2721-25KGBL
SPCMA2721-50KGBL
SPCMA2721-500GM
SPCMA2721-2.5KG
Allantoin 98,5 USP
Allantoin
Allantoin, USP is a moisturising ingredient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₄H₆N₄O₃ Molecular Weight: 158.12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005260 CAS-Nummer: 97-59-6 |
Spezifikation Testergebnisse
Assay | 98.5 - 101.0% |
Acidity or Alkalinity | To Pass Test |
Loss On Drying | max. 0.1 % |
Organic Impurities | max. 0.5 % |
Residue On Ignition (Sulphated Ash) | max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...