Sunflower seed oil

Lieferant: Spectrum Chemical

Synonyme: Helianthus Annuus (Sunflower) Seed Oil

S1929-4LTGL S1929-500MLGL
SPCMS1929-4LTGLEA 523 EUR
SPCMS1929-4LTGL SPCMS1929-500MLGL
Sunflower seed oil
Sunflower seed oil

Sunflower Seed Oil is, as its name suggests, the oil of sunflower seeds that is extracted by the compression of the seeds. This non-volatile oil is most commonly used in food and as a frying oil. It is also used in cosmetics as an emollient. It is also mixed with diesel to run diesel combustion engines.

Dichte: 0,90
Lagertemperatur: Raumtemperatur
MDL: MFCD00132403
CAS-Nummer: 8001-21-6

Bestellen

Spezifikation Testergebnisse

Synonyms Helianthus Annuus (Sunflower) Seed Oil
Free Fatty Acids APPROX. 0.13 %
Iodine Value Min. 110 - 143
Saponification Value Min. 188 - 194
Peroxide Value Max. 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR