Thiamine nitrate ≥98,0% USP

Lieferant: Spectrum Chemical

Synonyme: Vitamin B1 Nitrate

TH117-1KG TH117-25KGBL TH117-100GM
SPCMTH117-1KGEA 1140 EUR
SPCMTH117-1KG SPCMTH117-25KGBL SPCMTH117-100GM
Thiamine nitrate ≥98,0% USP
Thiamine nitrate

Thiamine Mononitrate, USP is the mononitrate salt form of a material also known as vitamin B1.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₁₂H₁₇N₅O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00036330
CAS-Nummer: 532-43-4

Bestellen

Spezifikation Testergebnisse

Assay (Dried Basis) 98.0 - 102.0%
pH of a 2% Solution @ 25°C 6.0 - 7.5
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.2 %
Chloride (Cl) max. 0.06 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR