Almond oil NF, sweet

Lieferant: Spectrum Chemical

AL215-200LTBL AL215-20LT AL215-4LTGL AL215-100ML
SPCMAL215-200LTBLEA 6490 EUR
SPCMAL215-200LTBL SPCMAL215-20LT SPCMAL215-4LTGL SPCMAL215-100ML
Almond oil NF, sweet
Almond oil

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Lagertemperatur: Raumtemperatur
MDL: MFCD00130467
CAS-Nummer: 8007-69-0

Bestellen

Spezifikation Testergebnisse

Specific Gravity @ 25°C 0.910 - 0.915
Fatty Acid Composition To Pass Test
Sterol Composition To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR