Vegetable oil brominated FCC
Lieferant: Spectrum Chemical
Brominated Vegetable Oil, FCC, also known as BVO, is a mix of plant-derived triglycerides and is used to help emulsify citrus flavored drinks. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
8016-94-2 |
Spezifikation Testergebnisse
Free Bromine | To Pass Test |
Specific Gravity | 1.325-1.335 |
Free Fatty Acids (As Oleic Acid) | max. 2.5 % |
Iodine Value | max. 16 |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...