DURAN® Volumetric Flasks with Stopper Bed
Lieferant: DWK Life Sciences
213725479 213722473 213722576 213723272 213723675 213724474 213726372 213726878 213727377 213728673
SCOT213725479EA 448 EUR
SCOT213725479 SCOT213722473 SCOT213722576 SCOT213723272 SCOT213723675 SCOT213724474 SCOT213726372 SCOT213726878 SCOT213727377 SCOT213728673
DURAN® Volumetric Flasks with Stopper Bed
Kolben Messkolben
Without graduation mark, imprint and stopper.
  • Manufactured from borosilicate 3.3 glass
  • With pre-ground plug base
  • Suitable for DIN EN ISO 1042
  • Uncalibrated glass flasks without calibration mark and imprint
Bestellinformation: Supplied without stoppers.