BAM 7 ≥98%

Lieferant: Cayman Chemical

Synonyme: 3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7

14566-1 14566-10 14566-25 14566-5
CAYM14566-1EA 43 EUR
CAYM14566-1 CAYM14566-10 CAYM14566-25 CAYM14566-5
BAM 7 ≥98%
BAM 7
Formel: C₂₁H₁₉N₅O₂S
Molecular Weight: 405,476 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01246556
CAS-Nummer: 331244-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR