MZone™ FlatMop™ Slip-On Mop Covers
Lieferant: MICRONOVA
MZSM11-16S MZSM11-16SIR MZSM11-9SIR MZSM56-14SIR MZSM56-18S MZSM56-18SIR MZSM620-14SIR MZSM11-14SIR
MICRMZSM11-16SEA 575 EUR
MICRMZSM11-16S MICRMZSM11-16SIR MICRMZSM11-9SIR MICRMZSM56-14SIR MICRMZSM56-18S MICRMZSM56-18SIR MICRMZSM620-14SIR MICRMZSM11-14SIR
MZone™ FlatMop™ Slip-On Mop Covers
Mopps
These slip-on, tab removal flat head mop covers are ideal for cleaning and sanitizing all surfaces and equipment in the cleanroom. A choice of fabric options provide excellent absorbency, effective cleaning, and disinfectant dispersal for specialized cleaning.
  • Easy to change covers fit securely over mop frame
  • Urethane foam interiors fully encased in a choice of fabric:
  • NovaPoly - 100% smooth knit polyester provides excellent disinfectant dispersal
  • PolySorb - 100% textured polyester for removing residues and dispersing disinfectant
  • MegaTex - 100% non-woven polyamide is absorbent with a mildly abrasive surface
  • Microfiber - 80% polyester or 20% nylon for maximum absorbency and residue removal
  • PolyMesh - 100% polyester with an abrasive mesh layer for exellent residue removal
  • The 228,6 mm covers are excellent for pass-throughs and smaller spaces
  • The 355,6 and 457,2 mm covers are ideal for floors, walls, and ceilings
The Micronova MZone™ FlatMop™ mop covers fit onto the 360° swivel SlimLine™ adapter, which can be locked at 180° for more controlled cleaning. Available gamma irradiated for aseptic environments. Suitable for use with strong disinfectants and repeated autoclaving.
Connection: Slip-on, tab removal
Interior: Urethane foam