Hemin chloride (aus Rind) ≥90%, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride
M-Clarity™ Qualitätsstufe = MQ200
H9039-100G
H9039-10G
H9039-1G
SIALH9039-100GEA
860
EUR
SIALH9039-100G
SIALH9039-10G
SIALH9039-1G
Hemin chloride (aus Rind) ≥90%, Sigma-Aldrich®
Hemin chloride
Formel:
C₃₄H₃₂ClFeN₄O₄ Molecular Weight: 651,95 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00010726 CAS-Nummer: 16009-13-5 EINECS: 240-140-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...