X-Wipes Dispenser System
Lieferant: BODE CHEMIE
981442 981479
BODE981442EA 97.8 EUR
BODE981442 BODE981479
X-Wipes Dispenser System
Boxen Spenderbehälter
It is versatile helper, that ensures maximum compliance in high risk areas with simultaneously convenient and easy use.
  • patented system solution: optimal active substance release during disinfection
  • use-solution usable for 28 days
  • 2,5 L use solution of the needed disinfectant
  • Absorbent polyester fleece 60 gr/m²
  • Size 380×200 mm per wipe
Areas of application
BODE X-Wipes Safety Pack is recommended for the disinfecting surface cleaning in all areas of health care facilities and laboratories that require maximum hygiene, cost-effectivenessand user convenience, for example:
• on work surfaces
• on patient stretchers
• on near-patient surfaces
• on surfaces in isolation rooms
• to prevent cross contamination in critical hygiene areas such as Intensive Care Unit (ICU), Haematooncology, Neonatology,
Intensive Care Burn Unit
• in areas that have no water connection
Compatibility with the BODE X-Wipes Safety Pack can be used with all liquid HARTMANN surface disinfectants as well as for following instrument disinfectants: Bodedex® forte (b), Bomix® plus (a), Korsolex ® basic (a), Korsolex ® extra (a).
Bitte beachten Use disinfectants safely. Always read the label and product information before use.