PharmaFluor® FEP Tubings
Lieferant: Saint Gobain Life Sciences
PHFE6012503150 PHFE6050006250
GESSPHFE6012503150EA 115 EUR
GESSPHFE6012503150 GESSPHFE6050006250
PharmaFluor® FEP Tubings
Schläuche Transferschlauch
PharmaFluor® FEP fluoropolymer tubing meets the highest demands of biopharmaceutical, medical, and laboratory applications. This tubing is formulated to resist the absorption/adsorption of aqueous fluids, which minimises the risk of fluid alteration in single- or repeat- use applications. PharmaFluor FEP has outstanding chemical and physical characteristics and can be used in a broad range of temperatures, from −73 to +204 °C (−100 to +400 °F).
  • Chemically inert
  • Ultra-pure
  • Documented biocompatibility
  • Fully characterised
  • Extremely low sorption characteristics
  • Fully sterilisable via autoclave or gas
  • Excellent barrier with very low permeability
  • Autoclavable
PharmaFluor® FEP tubing has outstanding chemical and physical characteristics, and processing of PharmaFluor® FEP occurs under stringently controlled conditions. In addition to being chemically inert, this tubing can be used at temperatures ranging from −73 to +204 °C (−100 to +400 °F). It is also virtually unaffected by chemical sanitisers and cleaners and can be cleaned repeatedly without decreasing its service life. It is ideal for applications such as sterile filling and dispensing systems, insulin production, diagnostic equipment, general laboratory use, cell and tissue culture transport, analytical and process equipment in the biotechnology, pharmaceutical and environmental fields.
Hardness: 55 (shore D)
Standards: USP Class VI criteria, ISO 10993 and European Pharmacopoeia 3.1.9.