BG-NH2 für SNAP-Tag-Fusion
Lieferant: AAT BIOQUEST
Achtung
Synonyme: O6-(4-Aminomethyl-benzyl)guanine, 6-((4-(Aminomethyl)benzyl)oxy)-7H-purin-2-amine
12560 12561
AATB12560EA 307 EUR
AATB12560 AATB12561
BG-NH2 für SNAP-Tag-Fusion
BG-NH2
BG-NH2 is a carboxy-reactive building block for preparing of SNAP substrates from the NHS esters or other amine-reactive forms of labels or biological targets.
  • SNAP substrate building block
  • Carboxy-reactive
Formel: C₁₃H₁₄N₆O
Molecular Weight: 270,2899 g/mol
MDL: MFCD22577247
CAS-Nummer: 674799-96-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb86cd6e66e8b46395e45ac0cbac7f764c00287f&vid=f1d7ef50bbe7ac933853de2c6e3fb2c97f98dc03&prid=web&clid=SAPDAM