FITC-C6-DEVD-FMK
Lieferant: AAT BIOQUEST
Achtung
13406 13408
AATB13406EA 769 EUR
AATB13406 AATB13408
FITC-C6-DEVD-FMK
FITC-C6-DEVD-FMK
Activation of caspases plays a central role in apoptosis.
  • Caspase detection
  • Minimal cytotoxicity
  • Cell permeable
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fefdb03b705d5b84fe1e19c2edf94a1aa3b5ea4&vid=ba81d28567b5379cc675d15e95a3b022ae97bacc&prid=web&clid=SAPDAM