Dihydrorhodamine 123 ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate, 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester
15206
15207
AATB15206EA
152
EUR
AATB15206
AATB15207
Dihydrorhodamine 123 ≥95%
Dihydrorhodamine 123
DihydrorH₂O₂damine 123 is by far the most-used probe for measurement of intracellular H₂O₂.
Formel:
C₂₁H₁₈N₂O₃ Molecular Weight: 346.386 g/mol |
MDL:
MFCD04221428 CAS-Nummer: 109244-58-8 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...