DAF-FM diacetate zur Stickoxiderkennung
Lieferant: AAT BIOQUEST
Achtung
Synonyme: DAF-FM DA, 4-Amino-5-methylamino-2',7'-difluorofluorescein diacetate
16298 16299
AATB16298EA 292 EUR
AATB16298 AATB16299
DAF-FM diacetate zur Stickoxiderkennung
DAF-FM diacetate
DAF-FM diacetate is a more cell-permeable version of DAF-FM, a sensitive fluorescent indicator for the detection and bioimaging of nitric oxide (NO).
  • Sensitive NO detection
Formel: C₂₅H₁₈F₂N₂O₇
Molecular Weight: 496,4 g/mol
CAS-Nummer: 254109-22-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed88a9756cd3f37701978eb6d7fb03bd562996e6&vid=1bf04281a70aaaa0cb288768f31ac829cb5a57fc&prid=web&clid=SAPDAM