Calbryte™ 630 AM
Lieferant: AAT BIOQUEST
|
Achtung
|
20720
20721
20722
AATB20720EA
307
EUR
AATB20720
AATB20721
AATB20722
Calbryte™ 630 AM
Calbryte™ 630 AM
Calcium measurement is critical for numerous biological investigations.
- Improved background/signal ratio
- Excellent intracellular retention
- Long-wavelength emission
- Cell permeant
Bitte beachten For research use only.
Molecular Weight:
1234.84 g/mol |
ADR:
5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...