FLUO 3/AM ≥95%, Ultrareine Qualität
Lieferant: AAT BIOQUEST
Achtung
Synonyme: 4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester
21011 21013
AATB21011EA 273 EUR
AATB21011 AATB21013
FLUO 3/AM ≥95%, Ultrareine Qualität
FLUO 3/AM
Calcium measurement is critical for numerous biological investigations.
Formel: C₅₁H₅₀Cl₂N₂O₂₃
Molecular Weight: 1129.85 g/mol
CAS-Nummer: 121714-22-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f55b1d92dba1e89085dbe68660387d394272563&vid=840b718657425db88be3b300370ce6054867237b&prid=web&clid=SAPDAM