Cal-520® sodium salt
Lieferant: AAT Bioquest
Achtung
21135 21136
AATB21135EA 460 EUR
AATB21135 AATB21136
Cal-520® sodium salt
Cal-520® sodium salt
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell impermeant
Bitte beachten For research use only.
ADR: 5,II