N-Boc-ethylendiamin

Lieferant: AAT BIOQUEST
Achtung

Synonyme: t-boc-N-EDA

4013 4014
AATB4013EA 614 EUR
AATB4013 AATB4014
N-Boc-ethylendiamin
N-Boc-ethylendiamin

N-BOC-ethylenediamine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.

Formel: C₇H₁₆N₂O₂
MDL: MFCD00191871
CAS-Nummer: 57260-73-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71ac086016765050fcc47612391a79da06f250c2&vid=c23f011e0effc002f6ac0fc6652ba240d688a47a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR