Natriumhydrogencarbonat-[13C], Sigma-Aldrich®

Lieferant: Merck

Synonyme: (13C)-Natriumbicarbonat, (13C)Natriumbicarbonat

M-Clarity™ Qualitätsstufe = MQ200

372382-1G 372382-250MG
SIAL372382-1GEA 238 EUR
SIAL372382-1G SIAL372382-250MG
Natriumhydrogencarbonat-[13C], Sigma-Aldrich®
Natriumhydrogencarbonat-[13C]
Formel: NaHCO₃
Molecular Weight: 85 g/mol
CAS-Nummer: 87081-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR