ABTS® Peroxidase Substrate Kit, 1-Component Microwell
Lieferant: SERACARE LIFE SCIENCES
5120-0041 5120-0046
KPLI5120-0041EA 880 EUR
KPLI5120-0041 KPLI5120-0046
ABTS® Peroxidase Substrate Kit, 1-Component Microwell
Tests Enzymtests
KPL ABTS 1-component microwell peroxidase substrate develops a blue-green product when reacted with horseradish peroxidase labeled conjugates in microwell plates or tubes. The expected appearance of
the solution is colorless to light green. Acceptable background absorbance of unreacted substrate is ≤0,025, as measured at 405 nm. KPL ABTS 1-component microwell peroxidase substrate is not suitable for membrane or immunohistochemical staining.