iFluor® 647, Succinimidyl Ester
Lieferant: AAT BIOQUEST
1031
71031
71510
71560
AATB1031EA
307
EUR
AATB1031
AATB71031
AATB71510
AATB71560
iFluor® 647, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- pH-Independent Signal
Bitte beachten For research use only.