Fluorescein diphosphate tetraammonium salt (FDP) ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Fluorescein (tetraammonium)
11600
11601
AATB11600EA
564
EUR
AATB11600
AATB11601
Fluorescein diphosphate tetraammonium salt (FDP) ≥95%
Fluorescein diphosphate tetraammonium salt (FDP)
Upon interaction with phosphatases the colourless and non-fluorescent FDP is hydrolysed to highly fluorescent fluorescein, which exhibits excellent spectral properties that match the optimal detection window of most fluorescence instruments that are equipped with the Argon laser excitation.
- Useful for multiple applications
Formel:
C₂₀H₂₆N₄O₁₁P₂ Molecular Weight: 560.393 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00278758 CAS-Nummer: 217305-49-2 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...